missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PAUF/ZG16B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7777
This item is not returnable.
View return policy
Description
PAUF/ZG16B Monoclonal specifically detects PAUF/ZG16B in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PAUF/ZG16B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q96DA0 | |
| ZG16B | |
| E. coli-derived recombinant human PAUF/ZG16B Gly53-Arg208 Accession # Q96DA0 | |
| 100 μg | |
| Primary | |
| Detects human PAUF/ZG16B in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 817310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp762P2216, EC 3.1.2, EC 3.4.19.12, EECP, HRPE773, hTrabid, JCLN2, LOC124220, PRO1567, TRABIDubiquitin thioesterase ZRANB1, TRAF-binding protein domain, ZG16B, Zinc finger Ran-binding domain-containing protein 1, zinc finger, RAN-binding domain containing 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 124220 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction