missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PAWR Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6885
This item is not returnable.
View return policy
Description
PAWR Polyclonal specifically detects PAWR in Human samples. It is validated for Immunocytochemistry.
Specifications
| PAWR | |
| Polyclonal | |
| Unconjugated | |
| CAD88640 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5074 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 5-15 ug/mL | |
| PAWR | |
| E. coli-derived recombinant human PAWR Arg2-Ala121 Accession # CAD88640 | |
| 100 μg | |
| Primary | |
| Detects human PAWR in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction