missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
207.00 € - 507.00 €
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | 2706A |
| Dilution | Immunohistochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700857
![]() |
R&D Systems
MAB108581-SP |
25 μg |
207.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752977
![]() |
R&D Systems
MAB108581-100 |
100 μg |
507.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| PD-1 | |
| Immunohistochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5133 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2706A | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Human PD-1 synthetic peptide | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title