missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-L2/B7-DC Antibody, R&D Systems™
Mouse Monoclonal Antibody
207.00 € - 599.00 €
Specifications
| Antigen | PD-L2/B7-DC/PDCD1LG2 |
|---|---|
| Clone | 1054620 |
| Dilution | Blockade of Receptor-ligand Interaction |
| Applications | Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18110931
|
R&D Systems
MAB11351-SP |
25 μg |
207.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18122081
|
R&D Systems
MAB11351-100 |
100 μg |
599.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Blocking AssaySpecifications
| PD-L2/B7-DC/PDCD1LG2 | |
| Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
| Human embryonic kidney cell, HEK293-derived recombinant human PD-L2/B7-DC, Accession # Q9BQ51 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1054620 | |
| Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BQ51 | |
| 80380 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title