Learn More
Human PDGF R alpha (Research Grade Olaratumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE
Human Monoclonal Antibody
Brand:  R&D Systems MAB10352-100
 View more versions of this product
Product Code. 18794587
  • 518.00 € / 100µg
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDGFR alpha
Monoclonal
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Human
Protein A or G purified from cell culture supernatant
RUO
5156
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG1
Flow Cytometry, CyTOF
Hu136
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Human PDGF R alpha
100 μg
Primary
Detects Human PDGF R alpha in direct ELISAs.This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Olaratumab. This product is for research use only.
Human
Purified