missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pentraxin 3/TSG-14 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB18261-SP
This item is not returnable.
View return policy
Description
Pentraxin 3/TSG-14 Monoclonal specifically detects Pentraxin 3/TSG-14 in Human samples. It is validated for ELISA.
Specifications
| Pentraxin 3/TSG-14 | |
| Monoclonal | |
| Unconjugated | |
| P26022 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5806 | |
| Human | |
| Purified |
| ELISA | |
| 247936 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 | |
| Mouse myeloma cell line NS0-derived recombinant human Pentraxin 3/TSG-14, Glu18-Ser381, Accession # P26022 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction