missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Perforin Antibody, R&D Systems™

Mouse Monoclonal Antibody
196.00 € - 458.00 €
Specifications
| Antigen | Perforin |
|---|---|
| Clone | 911710 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731847
![]() |
R&D Systems
MAB10385-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18774597
![]() |
R&D Systems
MAB10385-100 |
100 μg |
458.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for ELISA.Specifications
| Perforin | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P14222 | |
| 5551 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 911710 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1 | |
| E. coli-derived human Perforin, Pro22-Trp555, Accession # P14222 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title