missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-CEP250/CNAP1 (S2421) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11339-100
This item is not returnable.
View return policy
Description
CEP250 Monoclonal antibody specifically detects CEP250 in Human samples. It is validated for Western Blot
Specifications
| CEP250 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 250-KD, centrosomal Nek2-associated protein 1,250 kDa centrosomal protein, centrosomal protein 250kDa, centrosome associated protein, centrosome-associated protein CEP250, CEP2, CEP250, CNAP1, C-NAP1, MGC88542 | |
| Phosphopeptide containing the human CEP250/CNAP1 S2421 site, Accession # Q9BV73 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Western Blot | |
| 2739C | |
| Western Blot 1 μg/mL | |
| Q9BV73 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 11190 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction