Learn More
Human Phospho-Chk2 (T68) Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB1626-100
 View more versions of this product
Product Code. 18774889
  • 478.00 € / 100µg
Description
Chk2 Monoclonal specifically detects Chk2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chk2
Monoclonal
Unconjugated
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog
Rabbit
Protein A or G purified from cell culture supernatant
RUO
11200
Human
Purified
Western Blot, Immunocytochemistry
1238H
Western Blot 2 ug/mL, Immunocytochemistry 2-25 ug/mL
O96017
CHEK2
E.coli-derived recombinant human Phospho-Chk2 (T68) Accession # O96017
100 μg
Primary
Detects human Chk2 when phosphorylated at T68 in direct ELISAs and Western blots.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.