missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-GSK-3 beta (S9) Antibody, R&D Systems™

Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
Specifications
| Antigen | GSK-3 beta |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1253C |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide |
| Gene Accession No. | P49840 |
| Gene Alias | EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform |
| Gene Symbols | GSK3B |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?