missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Histone H2AX (S139) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
208.00 € - 451.00 €
Specifications
| Antigen | Histone H2AX |
|---|---|
| Clone | 2207D |
| Dilution | Western Blot 0.25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734679
![]() |
R&D Systems
MAB2288-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712298
![]() |
R&D Systems
MAB2288-100 |
100 μg |
451.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2AX Monoclonal specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| Histone H2AX | |
| Western Blot 0.25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P16104 | |
| 3014 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2207D | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| H2A.X, H2A/X, H2AFX | |
| Phospho-peptide containing human Histone H2AX S139 site Accession # P16104 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Histone H2AX when phosphorylated at S139 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title