Learn More
Human Phospho-MLKL (T357) Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB9187-100
 View more versions of this product
Product Code. 18734128
  • 508.00 € / 100µg
Description
MLKL Monoclonal specifically detects MLKL in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MLKL
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein
Phosphopeptide containing the human MLKL T357 site Accession # Q8NB16
100 μg
Primary
Detects human MLKL when phosphorylated at T357.
Human
Purified
Immunohistochemistry
954702
Immunohistochemistry 1-25 ug/mL
Q8NB16
Mouse
Protein A or G purified from cell culture supernatant
RUO
197259
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a