missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-p38 alpha (T180/Y182) Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB8692-SP
This item is not returnable.
View return policy
Description
p38 alpha Monoclonal specifically detects p38 alpha in Human samples. It is validated for Immunocytochemistry.
Specifications
| p38 alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MAPK14 | |
| Phosphopeptide containing human p38 alpha T180/Y182 site Accession # Q16539 | |
| 25 μg | |
| Primary | |
| Detects human p38 alpha when phosphorylated at T180/Y182. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 948243 | |
| Immunocytochemistry 5-25 ug/mL | |
| Q16539 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1432 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction