missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-p38 alpha (T180/Y182) Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB8692
This item is not returnable.
View return policy
Description
p38 alpha Monoclonal specifically detects p38 alpha in Human samples. It is validated for Immunocytochemistry.
Specifications
| p38 alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MAPK14 | |
| Phosphopeptide containing human p38 alpha T180/Y182 site Accession # Q16539 | |
| 100 μg | |
| Primary | |
| Detects human p38 alpha when phosphorylated at T180/Y182. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 948243 | |
| Immunocytochemistry 5-25 ug/mL | |
| Q16539 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1432 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur