missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-Rad17 (S635) Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18779438
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18779438 25 μg 25µg
18782568 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18779438 Supplier R&D Systems Supplier No. MAB1374SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Rad17 Monoclonal specifically detects Rad17 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rad17
Applications Western Blot
Classification Monoclonal
Clone 1244A
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. O75943
Gene Alias CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog
Gene Symbols RAD17
Host Species Rabbit
Immunogen Phosphopeptide containing the human Rad17 (S635) Accession # O75943
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5884
Test Specificity Detects human Rad17 when phosphorylated at S635 in Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.