missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Ribosomal Protein S6/RPS6 (S235/S236) Antibody, R&D Systems™

Description
Ribosomal Protein S6/RPS6 Monoclonal specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
Specifications
| Antigen | Ribosomal Protein S6/RPS6 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 973210 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P62753 |
| Gene Alias | Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6 |
| Host Species | Rat |
| Show More |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?