missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT3 (Y705) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4607-SP
This item is not returnable.
View return policy
Description
STAT3 Polyclonal specifically detects STAT3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| STAT3 (p tyr705) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| STAT3 | |
| Phosphopeptide containing human STAT3 Y705 site | |
| 25 μg | |
| Primary | |
| Detects human STAT3 when phosphorylated at Y705. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.25-0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6774 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction