missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-Vav-1 (Y160) Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18720038
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18720038 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18720038 Supplier R&D Systems Supplier No. MAB37861SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vav-1 Monoclonal specifically detects Vav-1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vav-1 (p Tyr160)
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 751411
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias oncogene vav, proto-oncogene vav, vav 1 guanine nucleotide exchange factor, vav proto-oncogene, Vav1, vav-T, VAVvav 1 oncogene
Gene Symbols VAV1
Host Species Mouse
Immunogen Phosphopeptide containing the human Vav-1 Y160 site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7409
Test Specificity Detects human VAV-1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.