missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PlGF-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
176.00 € - 407.00 €
Specifications
| Antigen | PlGF-4 |
|---|---|
| Clone | 1039228 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784197
![]() |
R&D Systems
MAB2644-SP |
25 μg |
176.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732677
![]() |
R&D Systems
MAB2644-100 |
100 μg |
407.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PlGF-4 Monoclonal antibody specifically detects PlGF-4 in Human samples. It is validated for ImmunocytochemistrySpecifications
| PlGF-4 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5228 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1039228 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P49763-4 | |
| Human embryonic kidney cell HEK293-derived human PlGF-4, Leu19-Arg242, Accession # P49763-4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title