missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PLUNC Antibody, R&D Systems™

Mouse Monoclonal Antibody
229.00 € - 522.00 €
Specifications
| Antigen | PLUNC |
|---|---|
| Clone | 251511R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751187
![]() |
R&D Systems
MAB18971-SP |
25 μg |
229.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792127
![]() |
R&D Systems
MAB18971-100 |
100 μg |
522.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLUNC Monoclonal specifically detects PLUNC in Human samples. It is validated for ELISA.Specifications
| PLUNC | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl | |
| E. coli-derived recombinant human PLUNC, Gln20-Val256, Accession No. Q9NP55 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PLUNC in direct ELISAs. |
| 251511R | |
| ELISA | |
| Unconjugated | |
| Human | |
| Q9NP55 | |
| 51297.0 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title