missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PMEL Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11439-SP
This item is not returnable.
View return policy
Description
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human samples. It is validated for Immunohistochemistry
Specifications
| PMEL17/SILV | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| D12S53EP1, gp100, ME20, ME20M, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17, PMEL17P100, premelanosome proteinME20M, SI, SIL, SILV, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| Chinese Hamster Ovary cell line, CHO-derived human PMEL, Lys25-Gln595, Accession # P40967 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 1067144 | |
| Immunohistochemistry 5-15 ug/mL | |
| P40967 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6490 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction