missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate Chitinase 3-like 1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB25991-500
This item is not returnable.
View return policy
Description
Chitinase 3-like 1 Monoclonal specifically detects Chitinase 3-like 1 in Human, Primate samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Chitinase 3-like 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1116 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 384327 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P36222 | |
| CHI3L1 | |
| Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 1 Tyr22-Thr383 Accession # P36222 | |
| 500 μg | |
| Primary | |
| Detects human and primate Chitinase 3-like 1 in direct ELISAs. | |
| Human, Primate | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction