missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate Cripto-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2772-500
This item is not returnable.
View return policy
Description
CRIPTO Monoclonal specifically detects CRIPTO in Human, Primate samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| CRIPTO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6997 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, Blocking Assay, Immunocytochemistry, CyTOF, ELISA | |
| 89633 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 0.1-0.5 ug/mL, Immunocytochemistry 5-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P13385 | |
| TDGF1 | |
| E. coli-derived recombinant human Cripto-1 Arg38-Tyr188 Accession # P13385 | |
| 500 μg | |
| Primary | |
| Detects human and primate Cripto-1 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse Cripto-1, recombinant human (rh) EGF, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 2, rhTGF-beta 3, or rhCryptic is observed. | |
| Human, Primate | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction