missing translation for 'onlineSavingsMsg'
Learn More

Human/Primate Cripto-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18793778
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18793778 25 μg 25µg
18722347 100 μg 100µg
18740827 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18793778 Supplier R&D Systems Supplier No. MAB2772SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRIPTO Monoclonal specifically detects CRIPTO in Human, Primate samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIPTO
Applications Flow Cytometry, Blocking Assay, Immunocytochemistry, CyTOF, ELISA
Classification Monoclonal
Clone 89633
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 0.1-0.5 ug/mL, Immunocytochemistry 5-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13385
Gene Alias CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1
Gene Symbols TDGF1
Host Species Mouse
Immunogen E. coli-derived recombinant human Cripto-1 Arg38-Tyr188 Accession # P13385
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6997
Test Specificity Detects human and primate Cripto-1 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse Cripto-1, recombinant human (rh) EGF, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 2, rhTGF-beta 3, or rhCryptic is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.