missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-17/IL-17A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 18 publications
213.00 € - 2160.00 €
Specifications
| Antigen | IL-17/IL-17A |
|---|---|
| Clone | 41809 |
| Dilution | Immunoprecipitation 3 ug/100 ug cell lysate, Neutralization 1-3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Immunoprecipitation, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750539
![]() |
R&D Systems
MAB317-SP |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18771299
![]() |
R&D Systems
MAB317-100 |
100 μg |
534.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795538
![]() |
R&D Systems
MAB317-500 |
500 μg |
2160.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human, Primate samples. It is validated for Immunohistochemistry, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| IL-17/IL-17A | |
| Immunoprecipitation 3 ug/100 ug cell lysate, Neutralization 1-3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Primate | |
| Q16552 | |
| 3605 | |
| E. coli-derived recombinant human IL-17 Ile20-Ala155 Accession # Q16552 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and primate IL-17 in direct ELISAs. In direct ELISAs, approximately 12% cross-reactivity with recombinant canine IL-17 is observed and 25%-50% reactivity with recombinant human (rh) IL-17A/IL-17F heterodimer is observed. No cross-reactivity with recombinant mouse IL-17, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, or rhIL-17F is observed. |
| 41809 | |
| Immunoprecipitation, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| IL17A | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title