missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
241.00 € - 2205.00 €
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | 5355 |
| Dilution | Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Immunocytochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729407
![]() |
R&D Systems
MAB602-SP |
25 μg |
241.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756409
![]() |
R&D Systems
MAB602-100 |
100 μg |
540.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753838
![]() |
R&D Systems
MAB602-500 |
500 μg |
2205.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-2 Monoclonal specifically detects IL-2 in Human, Primate samples. It is validated for Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
| IL-2 | |
| Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Primate | |
| NP_000577 | |
| 3558 | |
| E. coli-derived recombinant human IL-2 Ala21-Thr153 Accession # NP_000577 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and primate IL-2 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-3, rhIL-4, rhIL-6, rhIL-7, rhIL-8, recombinant mouse (rm) IL-1 beta , rmIL-2, rmIL-3, rmIL-4, rmIL-5, rmIL-6, or rmIL-7 is observed. |
| 5355 | |
| Immunocytochemistry, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| IL2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title