missing translation for 'onlineSavingsMsg'
Learn More

Human/Primate IL-6 Biotinylated Antibody, R&D Systems™

Product Code. 18061712
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18061712 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18061712 Supplier R&D Systems Supplier No. BAF206
  • ONLINE EXCLUSIVE PRICE
    777.56 €
    689.85 € / 50µg
    Save 87.71 €
    11% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 29 publications

IL-6 Polyclonal specifically detects IL-6 in Human∣Papio cynocephalus (Yellow Baboon)∣Primate samples. It is validated for Antibody Array Development∣Array Development∣Bioassay∣Capture Assay∣Electrochemiluminescent Assay∣ELISA Capture∣ELISA Detection∣ELISA Detection (Matched Antibody Pair)∣ELISA Development∣ELISA Development (Detection)∣Functional Assay∣Immunoassay Development∣Immunocytochemistry∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Array, Bioassay, Electrochemiluminescent Assay, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Development, ELISA Development (Detection), Functional Assay, Immunoassay, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P05231
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Gene Symbols IL6
Host Species Goat
Immunogen E. coli-derived recombinant human IL-6 Pro29-Met212 Accession # P05231
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3569
Test Specificity Detects human and primate IL-6 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant porcine IL-6 is observed and less than 0.01% cross-reactivity with recombinant mouse IL-6 and recombinant rat IL-6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Yellow Baboon, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.