missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro BDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB31752-100
This item is not returnable.
View return policy
Descripción
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for ELISA.
Especificaciones
| BDNF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF Met1-Arg247 Accession # P23560 | |
| 100 μg | |
| 627 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 584431 | |
| ELISA | |
| P23560 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Pro-BDNF in direct ELISAs and sandwich immunoassays. | |
| Human | |
| Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido