missing translation for 'onlineSavingsMsg'
Learn More

Human Pro MMP-13 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18731918
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731918 25 μg 25µg
18121041 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18731918 Supplier R&D Systems Supplier No. MAB913SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-13 Monoclonal specifically detects MMP-13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-13
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Affinity Purification
Classification Monoclonal
Clone 87518
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P45452
Gene Alias CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13
Gene Symbols MMP13
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human MMP-13 Leu20-Cys471 Accession # P45452
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4322
Test Specificity Detects human Pro MMP-13 in direct ELISAs and Western blots. Does not detect active recombinant human MMP-13. In Western blots, no cross-reactivity with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -12 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.