missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro MMP-13 Antibody, R&D Systems™

Description
MMP-13 Monoclonal specifically detects MMP-13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification.
Specifications
Specifications
| Antigen | MMP-13 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Affinity Purification |
| Classification | Monoclonal |
| Clone | 87518 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, ELISA, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P45452 |
| Gene Alias | CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13 |
| Gene Symbols | MMP13 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?