missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Procalcitonin Antibody, R&D Systems™

Mouse Monoclonal Antibody
208.00 € - 422.00 €
Specifications
| Antigen | Procalcitonin |
|---|---|
| Clone | 944002 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18709138
![]() |
R&D Systems
MAB83501-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744698
![]() |
R&D Systems
MAB83501-100 |
100 μg |
422.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| Procalcitonin | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P01258 | |
| 796 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 944002 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
| Human Procalcitonin synthetic peptide Ala26-Asp38 Accession # P01258 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Procalcitonin in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title