missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protein Disulfide Isomerase/P4HB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4236-SP
This item is not returnable.
View return policy
Description
Protein Disulfide Isomerase/P4HB Monoclonal specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| Protein Disulfide Isomerase/P4HB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1-2 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P07237 | |
| P4HB | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protein Disulfide Isomerase/P4HB Asp18-Lys505 Accession # P07237 | |
| 25 μg | |
| Primary | |
| Detects human Protein Disulfide Isomerase/P4HB in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 537331 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDI, PDIA1, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHB, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, Thbp, thyroid hormone-binding protein p55 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5034 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction