missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTGER3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
203.00 € - 447.00 €
Specifications
| Antigen | PTGER3 |
|---|---|
| Clone | 606609 |
| Dilution | Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780207
![]() |
R&D Systems
MAB10243-SP |
25 μg |
203.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770347
![]() |
R&D Systems
MAB10243-100 |
100 μg |
447.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTGER3 Monoclonal specifically detects PTGER3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| PTGER3 | |
| Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Lyophilized | |
| Human | |
| P43115 | |
| 5733 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 606609 | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EP3, EP3EP3e, EP3-I, EP3-II, EP3-III, EP3-IV, MGC141828, MGC141829, MGC27302, PGE receptor EP3 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP3 subtype, PGE2-R, prostaglandin E receotor EP3 subtype 3 isoform, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP3 receptor | |
| NS0 mouse myeloma cell line transfected with human PTGER3, Met1-Arg390, Accession # P43115 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PTGER3 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title