missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTH1R/PTHR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB57091-SP
This item is not returnable.
View return policy
Description
PTH1R/PTHR1 Monoclonal specifically detects PTH1R/PTHR1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| PTH1R/PTHR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHR, PTHR1, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor | |
| Chinese hamster ovary cell line CHO-derived recombinant human PTHR1 Tyr23-Met189 Accession # Q03431 | |
| 25 μg | |
| Primary | |
| Detects human PTHR1 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 542407 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q03431 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5745 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction