missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTP1B Antibody, R&D Systems™

Mouse Monoclonal Antibody
170.00 € - 422.00 €
Specifications
| Antigen | PTP1B/PTPN1 |
|---|---|
| Clone | 208607 |
| Dilution | Immunohistochemistry 1-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741577
![]() |
R&D Systems
MAB13662-SP |
25 μg |
170.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704697
![]() |
R&D Systems
MAB13662-100 |
100 μg |
422.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human samples. It is validated for .Specifications
| PTP1B/PTPN1 | |
| Immunohistochemistry 1-25 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P18031 | |
| 5770 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 208607 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, PTPN1, tyrosine-protein phosphatase non-receptor type 1 | |
| E. coli-derived recombinant human PTP1B, Glu2-Thr435, Accession # P18031 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PTP1B in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title