Learn More
Human RANK/TNFRSF11A Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB6832-SP
 View more versions of this product
Product Code. 18722687
  • 224.00 € / 25µg
Description
RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RANK/TNFRSF11A
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
RANK/TNFRSF11A
Mouse myeloma cell line NS0-derived recombinant human RANK/TNFRSF11A, extracellular domain, Accession # Q9Y6Q6
25 μg
Primary
Human
Purified
Immunohistochemistry, Immunocytochemistry
80710
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Q9Y6Q6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a