missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat CCR4 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems FAB1567F-025
This item is not returnable.
View return policy
Description
CCR4 Monoclonal specifically detects CCR4 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
| CCR4 | |
| Monoclonal | |
| Fluorescein | |
| P51679 | |
| CCR4 | |
| Human CCR4 transfectants Met1-Leu360 Accession # P51679 | |
| 25 Tests | |
| 1233 | |
| Human, Rat | |
| Purified |
| Flow Cytometry | |
| 205410 | |
| Flow Cytometry 10 uL/10^6 cells | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Stains human CCR4-transfected cell lines but not the corresponding parent cell lines. Also detects human CCR4 on human PBLs and platelets and rat CCR4 on rat splenocytes. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction