missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat CCR8 PerCP-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1429C-100
This item is not returnable.
View return policy
Description
CCR8 Monoclonal specifically detects CCR8 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
| CCR8 | |
| Monoclonal | |
| PerCP | |
| P51685 | |
| CCR8 | |
| BaF3 mouse pro-B cell line transfected with human CCR8 Met1-Leu355 Accession # P51685 | |
| 100 Tests | |
| 1237 | |
| Human, Rat | |
| Purified |
| Flow Cytometry | |
| 191704 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CCR8. Neutralizes the bioactivity of human CCR8 and specifically stains human CCR8-transfected cell lines but not the parent cell lines. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction