missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat MDGA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 427.00 €
Specifications
| Antigen | MDGA2 |
|---|---|
| Clone | 803732 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
18763609
![]() |
R&D Systems
MAB5184-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18749898
![]() |
R&D Systems
MAB5184-100 |
100 μg |
427.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
MDGA2 Monoclonal specifically detects MDGA2 in Human, Rat samples. It is validated for Flow Cytometry, CyTOF-ready.Specifikationer
| MDGA2 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Mouse | |
| Human, Rat | |
| Q72553 | |
| 161357 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 803732 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1 | |
| Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp934 Accession # Q7Z553 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MDGA2 in direct ELISAs. Detects rat MDGA2 in Flow Cytometry. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel