missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat Vinculin Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6896
This item is not returnable.
View return policy
Description
Vinculin Polyclonal specifically detects Vinculin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Vinculin | |
| Polyclonal | |
| Unconjugated | |
| P18206 | |
| VCL | |
| E. coli-derived recombinant human Vinculin Lys1020-Gln1134 Accession # P18206 | |
| 100 μg | |
| Primary | |
| Detects recombinant human Vinculin in direct ELISAs and human and rat Vinculin in Western blots. | |
| Human, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CMD1W, CMH15, Metavinculin, MVCL, VCL, vinculin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7414 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction