Learn More
Human RBFOX3/NeuN Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11665-100
 View more versions of this product
Product Code. 30484806
  • 572.00 € / 100µg
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human samples. It is validated for Multiplex Immunofluorescence,Western Blot,Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBFOX3/NeuN
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3
Synthetic Peptide, Accession # A6NFN3
100 μg
Primary
Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Multiplex Immunofluorescence, Western Blot, Immunohistochemistry
1094645
Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL
A6NFN3
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
146713
Human
Antibody
IgG1