missing translation for 'onlineSavingsMsg'
Learn More

Human RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

209.00 € - 1680.00 €

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Clone 393006
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18714268
GREENER_CHOICE
View Documents
R&D Systems
MAB33781-SP
25 μg
209.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18744498
GREENER_CHOICE
View Documents
R&D Systems
MAB33781-100
100 μg
425.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18792318
GREENER_CHOICE
View Documents
R&D Systems
MAB33781-500
500 μg
1680.00 €
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

RBP4/Retinol-Binding Protein 4
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P02753
5950
Mouse myeloma cell line NS0-derived recombinant human RBP4/Retinol-Binding Protein 4 Glu19-Leu201 Accession # P02753
Primary
Sterile PBS to a final concentration of 0.5 mg/mL.
Detects human RBP4/Retinol-Binding Protein 4 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse RBP4 is observed.
393006
ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma
RBP4
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.