missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human RBX1 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ MAB11499-SP
This item is not returnable.
View return policy
Description
RBX1 Monoclonal antibody specifically detects RBX1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Immunocytochemistry
Specifications
| RBX1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| BA554C12.1, EC 6.3.2.-, FLJ60363, MGC13357, Protein ZYP, Rbx1, Regulator of cullins 1, Ring Box 1, RING box protein 1, RING finger protein 75, ring-box 1, ring-box 1, E3 ubiquitin protein ligase, RING-box protein 1, RNF75, RNF75MGC1481, ROC1E3 ubiquitin-protein ligase RBX1, ZYP protein | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL1/RBX1 Complex, Met1-His108, Accession # P62877 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry | |
| 1076128 | |
| Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| P62877 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9978 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction