missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Rectum Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired)

Product Code. 18245635 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18245635 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18245635 Supplier Novus Biologicals™ Supplier No. NB82089078
  • ONLINE EXCLUSIVE PRICE
    1110.00 €
    1047.90 € / 1 sets
    Save 62.10 €
    5% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The Human Rectum Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Digestive/Waste
Quantity 1 Set
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Matched Pair - Total Protein - Human Primary Tumor and Normal Tissue (PP): Rectum
Cellular Fraction Whole
Tissue Condition Primary Tumor and Normal Paired
Cross Reactivity Human
Host Species Human
Concentration 5 mg/mL
Product Type Human Rectum Whole Tissue Lysate (Adult Primary Tumor and Normal Paired)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.