missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RIPK2/RIP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB103871-100
This item is not returnable.
View return policy
Description
RIPK2 Monoclonal specifically detects RIPK2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RIPK2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CARD3, CARD-containing IL-1 beta ICE-kinase, CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, CARDIAKCCK, EC 2.7.11, EC 2.7.11.1, GIG30, growth-inhibiting gene 30, receptor interacting protein 2, receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase, Receptor-interacting protein 2, receptor-interacting serine/threonine-protein kinase 2, receptor-interacting serine-threonine kinase 2, RICK, RICKGIG30, RIP2, RIP-2, RIP2CARD-carrying kinase, RIP-like-interacting CLARP kinase | |
| E. coli-derived human RIPK2/RIP2, Met1-Met540, Accession # O43353 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| 1018218 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 5-25 μg/mL | |
| O43353 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8767 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction