missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4570-SP
This item is not returnable.
View return policy
Description
S100A8 Polyclonal specifically detects S100A8 in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| S100A8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 60B8AG, CAGACP-10, calgranulin A, Calgranulin-A, Calprotectin L1L subunit, CFAG, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6279 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P05109 | |
| S100A8 | |
| E. coli-derived recombinant human S100A8 Met1-Glu93 Accession # P05109 | |
| 25 μg | |
| Primary | |
| Detects human S100A8 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant mouse S100A8, recombinant human (rh) S100A1, and rhS100P is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction