missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100B Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 461.00 €
Specifications
| Antigen | S100B |
|---|---|
| Clone | 1046234 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710077
![]() |
R&D Systems
MAB18203-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791507
![]() |
R&D Systems
MAB18203-100 |
100 μg |
461.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100B Monoclonal antibody specifically detects S100B in Human samples. It is validated for ImmunohistochemistrySpecifications
| S100B | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
| Human S100B synthetic peptide, Accession # P04271 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1046234 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04271 | |
| 6285 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title