missing translation for 'onlineSavingsMsg'
Learn More

Human SCF/c-kit Ligand Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 4 publications

208.00 € - 1710.00 €

Specifications

Antigen SCF/c-kit Ligand
Clone 13302
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18701389
GREENER_CHOICE
View Documents
R&D Systems
MAB655-SP
25 μg
208.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18712819
GREENER_CHOICE
View Documents
R&D Systems
MAB655-100
100 μg
436.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18783667
GREENER_CHOICE
View Documents
R&D Systems
MAB655-500
500 μg
1710.00 €
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SCF/c-kit Ligand Monoclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

SCF/c-kit Ligand
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P21583
4254
E. coli-derived recombinant human SCF Glu26-Ala189 Accession # P21583
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects recombinant human SCF in Western blots. In sandwich ELISAs, no significant cross-reactivity or interference was observed with 36 other cytokines.
13302
Western Blot, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
KITLG
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.