missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SEC13 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9055-100
This item is not returnable.
View return policy
Description
SEC13 Monoclonal specifically detects SEC13 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| SEC13 | |
| Monoclonal | |
| Unconjugated | |
| Supplied lyophilized from a solution in PBS and trehalose | |
| D3S1231E, D3S1231ESEC13L1npp-20, Npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13R, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae) | |
| E. coli-derived recombinant human SEC13 Gly123-Gln322 Accession # P55735 | |
| 100 μg | |
| Primary | |
| Detects human SEC13 in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1280A | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 0.3-25 ug/mL | |
| P55735 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6396 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction