Learn More
Human Serpin E2/PN1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB2980-SP
 View more versions of this product
Product Code. 18728829
  • 211.00 € / 25µg
Description
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serpin E2/PN1
Monoclonal
LYOPH
Western Blot 1 ug/mL
NP_001130000
SERPINE2
Mouse myeloma cell line NS0-derived recombinant human Serpin E2/PN1 Met1-Pro397 Accession # NP_001130000
25 μg
Primary
Detects human Serpin E2/PN1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A12, B6, B8, B9, C1, D1, F2, I1, recombinant mouse Serpin E2, or rhPEDF is observed.
Human
Purified
Western Blot
449888
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5270
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b